Novus Biologicals
Manufacturer Code:NBP305068100UL
Catalog # NB095228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 487-736 of human LEPREL2 (NP_055077.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B leprecan-like 2 P3H3 prolyl 3-hydroxylase 3; gene rich cluster, B; leprecan-like 2; Leprecan-like protein 2; procollagen-proline 3-dioxygenase; Prolyl 3-hydroxylase 3; Protein B
Gene Aliases: GRCB; HSU47926; LEPREL2; P3H3
UniProt ID: (Human) Q8IVL6
Entrez Gene ID: (Human) 10536
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.