Novus Biologicals
Manufacturer Code:NBP30501120UL
Catalog # NB094577
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 400-700 of human LEPREL1 (NP_060662.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ10718 leprecan-like 1 leprecan-like protein 1 MCVD myxoid liposarcoma associated protein 4 myxoid liposarcoma-associated protein 4 P3H2 prolyl 3-hydroxylase 2 prolyl 3-hydroxylase 3; leprecan-like 1; Leprecan-like protein 1; Myxoid liposarcoma-associated protein 4; procollagen-proline 3-dioxygenase 2; Prolyl 3-hydroxylase 2; prolyl 3-hydroxylase 3
Gene Aliases: LEPREL1; MCVD; MLAT4; P3H2
UniProt ID: (Human) Q8IVL5
Entrez Gene ID: (Human) 55214
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.