Invitrogen
Manufacturer Code:PA553527
Catalog # PIPA553527
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human LEPRE1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Growth suppressor 1; leprecan; Leprecan-1; leucine proline-enriched proteoglycan (leprecan) 1; Leucine- and proline-enriched proteoglycan 1; procollagen-proline 3-dioxygenase; Prolyl 3-hydroxylase 1; Prolyl 3-hydroxylase 1 Growth suppressor 1 Leucine- and proline-enriched proteoglycan 1 Leprecan-1 LEPRE1 GROS1 P3H1
Gene Aliases: GROS1; LEPRE1; OI8; P3H1; PSEC0109
UniProt ID: (Human) Q32P28
Entrez Gene ID: (Human) 64175
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.