Novus Biologicals
Manufacturer Code:NBP230460
Catalog # NBP230460
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: INKLEKAVAAAHTFFVGNPEHMEMQQNLDYYQTMSGVKEADFKDLETQPHMQEFRLGVRLYSEEQPQEAVPHLEAALQEYFVAYEECRALCEGPYDYDGYNYLEYNADLFQAITDHYIQVLNCKQNCVTELASHP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 1.14.11.7 GROS1OI8 Growth suppressor 1 leprecan Leprecan-1 Leucine- and proline-enriched proteoglycan 1 leucine proline-enriched proteoglycan (leprecan) 1 MGC117314 P3H1LEPRECAN prolyl 3-hydroxylase 1; Growth suppressor 1; leprecan; Leprecan-1; leucine proline-enriched proteoglycan (leprecan) 1; Leucine- and proline-enriched proteoglycan 1; procollagen-proline 3-dioxygenase; Prolyl 3-hydroxylase 1
Gene Aliases: GROS1; LEPRE1; OI8; P3H1; PSEC0109
UniProt ID: (Human) Q32P28
Entrez Gene ID: (Human) 64175
Molecular Function: extracellular matrix glycoprotein extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.