Abnova
Manufacturer Code:PAB8805
Catalog # 89-133-949
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids 25-208 of mouse Lepr. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant Lepr.
For Research Use Only
Protein Aliases: B219; CD295; db; diabetes; LEP-R; Leprb; LEPROT; Leptin receptor; leptin receptor gene-related protein; MGC105189; Modb1; OB receptor; OB-R; OB-RGRP; obese-like; obl; Obr
Gene Aliases: Db; diabetes; Lepr; Leprb; LEPROT; Modb1; OB-RGRP; obese-like; obl; Obr
UniProt ID: (Mouse) P48356
Entrez Gene ID: (Mouse) 16847
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.