Abnova
Manufacturer Code:PAB10167
Catalog # 89-114-675
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to mouse Lep. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C on dry atmosphere. After reconstitution with deionized water,store at 4°C at least six weeks. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant Lep.
For Research Use Only
Protein Aliases: Leptin; ob obese; Obesity factor
Gene Aliases: Lep; Ob; obese
UniProt ID: (Mouse) P41160
Entrez Gene ID: (Mouse) 16846
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.