Novus Biologicals
Manufacturer Code:NBP219346
Catalog # NBP219346
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human LEO1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Leo1 Paf1/RNA polymerase II complex component; Leo1 Paf1/RNA polymerase II complex component homolog (S. cerevisiae) RDLReplicative senescence down-regulated leo1-like protein RNA polymerase-associated protein LEO1; Leo1, Paf1/RNA polymerase II complex component, homolog; Replicative senescence down-regulated leo1-like protein; RNA polymerase-associated protein LEO1
Gene Aliases: LEO1; RDL
UniProt ID: (Human) Q8WVC0
Entrez Gene ID: (Human) 123169
Molecular Function:
DNA-directed RNA polymerase
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.