Novus Biologicals
Manufacturer Code:NBP271744IR
Catalog # NBP271744IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6C10 |
Immunogen | Full length recombinant protein of human LEMD3 (NP_055134) produced in HEK293T cells. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Inner nuclear membrane protein Man1; inner nuclear membrane protein Man1 integral inner nuclear membrane protein LEM domain containing 3 MAN1LEM domain-containing protein 3; integral inner nuclear membrane protein; LEM domain-containing protein 3
Gene Aliases: LEMD3; MAN1
UniProt ID: (Human) Q9Y2U8
Entrez Gene ID: (Human) 23592
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.