Novus Biologicals
Manufacturer Code:NBP266625
Catalog # NBP266625
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1C3 |
Immunogen | Recombinant full-length LEF-1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: DKFZp586H0919 FLJ46390 LEF-1 lymphoid enhancer-binding factor 1 T cell-specific transcription factor 1-alpha TCF1-alpha TCF1ALPHA; LEF-1; Lymphoid enhancer-binding factor 1; T cell-specific transcription factor 1-alpha; TCF1-alpha
Gene Aliases: LEF-1; LEF1; TCF10; TCF1ALPHA; TCF7L3
UniProt ID: (Human) Q9UJU2
Entrez Gene ID: (Human) 51176
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.