Invitrogen
Manufacturer Code:PA585555
Catalog # PIPA585555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the N-terminus region of human LEF1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LEF-1; LEF-1 Antibody lymphoid enhancer-binding factor 1 Antibody TCF10 Antibody TCF7L3 Antibody TCF1ALPHA Antibody; Lymphoid enhancer-binding factor 1; T cell-specific transcription factor 1-alpha; TCF1-alpha
Gene Aliases: LEF-1; LEF1; TCF10; TCF1ALPHA; TCF7L3
UniProt ID: (Human) Q9UJU2
Entrez Gene ID: (Human) 51176
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.