R&D Systems
Manufacturer Code:AF7647SP
Catalog # AF7647SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human LEF1 Met120-Met280 (predicted) Accession # Q9UJU2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp586H0919 FLJ46390 LEF-1 lymphoid enhancer-binding factor 1 T cell-specific transcription factor 1-alpha TCF10 TCF1-alpha TCF1ALPHA TCF7L3; LEF-1; Lymphoid enhancer-binding factor 1; T cell-specific transcription factor 1-alpha; TCF1-alpha
Gene Aliases: LEF-1; LEF1; TCF10; TCF1ALPHA; TCF7L3
UniProt ID: (Human) Q9UJU2
Entrez Gene ID: (Human) 51176
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.