Proteintech
Manufacturer Code:149721AP150UL
Catalog # 50-172-9876
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LEF1 Fusion Protein Ag6882 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LEF 1 LEF1 TCF1 alpha TCF1ALPHA; LEF-1; lymphoid enhancer binding factor 1 short isoform; Lymphoid enhancer-binding factor 1; T cell-specific transcription factor 1-alpha; TCF1-alpha
Gene Aliases: 3000002B05; AI451430; LEF-1; LEF1; TCF10; TCF1ALPHA; TCF7L3
UniProt ID: (Human) Q9UJU2, (Rat) Q9QXN1, (Mouse) P27782
Entrez Gene ID: (Human) 51176, (Rat) 161452, (Mouse) 16842
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.