R&D Systems
Manufacturer Code:AF3468
Catalog # AF3468
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human LEDGF Met1-Asn530 Accession # O75475 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CLL-associated antigen KW-7; CLL-associated antigen KW-7 Dense fine speckles 70 kDa protein DFS 70 DFS70 LEDGFDFS 70 Lens epithelium-derived growth factor MGC74712 p52 p75 PAIP PC4 and SFRS1 interacting protein 1 PC4 and SFRS1 interacting protein 2 PC4 and SFRS1-interacting protein PSIP1 PSIP2 transcriptional coactivator p52/p75 Transcriptional coactivator p75/p52; Dense fine speckles 70 kDa protein; DFS 70; Lens epithelium-derived growth factor; PC4 and SFRS1-interacting protein; transcriptional coactivator p52/p75; Transcriptional coactivator p75/p52
Gene Aliases: DFS70; LEDGF; p52; p75; PAIP; PSIP1; PSIP2
UniProt ID: (Human) O75475
Entrez Gene ID: (Human) 11168
Molecular Function:
growth factor
signaling molecule
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.