OriGene
Manufacturer Code:TA600147
Catalog # 50-167-7220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4B10 |
Immunogen | Full length human recombit protein of human LDLRAP1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ARH ARH1 ARH2 FHCB1 FHCB2; Autosomal recessive hypercholesterolemia protein; LDL receptor adaptor protein; Low density lipoprotein receptor adapter protein 1
Gene Aliases: ARH; ARH1; ARH2; FHCB1; FHCB2; LDLRAP1
UniProt ID: (Human) Q5SW96
Entrez Gene ID: (Human) 26119
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.