Invitrogen
Manufacturer Code:MA530637
Catalog # PIMA530637
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7G11G11E6 |
Immunogen | Recombit Human LDLR protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: LDL receptor; Low-density lipoprotein receptor; low-density lipoprotein receptor class A domain-containing protein 3
Gene Aliases: FH; FHC; LDLCQ2; LDLR
UniProt ID: (Human) P01130
Entrez Gene ID: (Human) 3949
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.