Novus Biologicals
Manufacturer Code:NBP106709
Catalog # NBP106709
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to an internal portion of the human LDL Receptor protein (within residues 500-550). [Swiss-Prot# P01130] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FH FHC LDL receptor LDLCQ2 low density lipoprotein receptor low-density lipoprotein receptor low-density lipoprotein receptor class A domain-containing protein 3; LDL receptor; Low-density lipoprotein receptor; low-density lipoprotein receptor class A domain-containing protein 3
Gene Aliases: FH; FHC; LDLCQ2; LDLR
UniProt ID: (Human) P01130
Entrez Gene ID: (Human) 3949
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.