Invitrogen
Manufacturer Code:PA544076
Catalog # PIPA544076
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human LDHC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cancer/testis antigen 32; CT32; L-lactate dehydrogenase C; L-lactate dehydrogenase C chain; lactate dehydrogenase 3, C chain, sperm specific; Lactate dehydrogenase C LDH3 LDHX MGC111073 CT32; lactate dehydrogenase c variant 1; lactate dehydrogenase C variant 3; lactate dehydrogenase c variant 4; lactate dehydrogenase c variant 5; lactate dehydrogenase C4; lactate dehydrogenase-X; LDH testis subunit; LDH-C; LDH-X; testis secretory sperm-binding protein Li 234P
Gene Aliases: CT32; Ldh-3; LDH-C; LDH-C4; LDH-X; LDH3; LDHC; Ldhc4; LDHX
UniProt ID: (Pig) Q9TSX5, (Human) P07864, (Mouse) P00342
Entrez Gene ID: (Bovine) 537256, (Pig) 100502559, (Dog) 476883, (Human) 3948, (Rat) 29634, (Mouse) 16833
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.