Invitrogen
Manufacturer Code:PA518780
Catalog # PIPA518780
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (DSDKEHWKNVHKQ) corresponding to the internal amino acids of LDHC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: L-lactate dehydrogenase C chain; lactate dehydrogenase 3, C chain, sperm specific; lactate dehydrogenase-X; LDH testis subunit; LDH-C; LDH-X; LDHC MGC111073 lactate dehydrogenase C LDH3 LDHX
Gene Aliases: Ldh-3; LDH-C4; Ldh-x; Ldh3; Ldhc; Ldhc4
UniProt ID: (Mouse) P00342
Entrez Gene ID: (Mouse) 16833
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.