Abnova
Manufacturer Code:PAB28316
Catalog # 89-129-820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of recombinant LDHC. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant LDHC.
For Research Use Only
Protein Aliases: Cancer/testis antigen 32; CT32; L-lactate dehydrogenase C chain; lactate dehydrogenase c variant 1; lactate dehydrogenase c variant 3; lactate dehydrogenase c variant 4; lactate dehydrogenase c variant 5; lactate dehydrogenase C4; LDH testis subunit; LDH-C; LDH-X; LDH3; LDHX; MGC111073; testis secretory sperm-binding protein Li 234P
Gene Aliases: CT32; LDH3; LDHC; LDHX
UniProt ID: (Human) P07864
Entrez Gene ID: (Human) 3948
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.