Invitrogen
Manufacturer Code:PA543141
Catalog # PIPA543141
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human LDHB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 281; H(B)-type lactate dehydrogenase; L-lactate dehydrogenase B chain; Lactate dehydrogenase B LDH-H TRG-5 LDH-B LDHBD; lactate dehydrogenase H chain; lactate dehydrogenase-H; lactate dehydrogenase-H (, EC 1.1.1.27); Lactate dehydrogenease B; LDH heart subunit; LDH-B; LDH-H; Renal carcinoma antigen NY-REN-46; testicular secretory protein Li 25
Gene Aliases: AI790582; H-Ldh; HEL-S-281; Ldh-2; LDH-B; LDH-H; Ldh2; LDHB; LDHBD; TRG-5
UniProt ID: (Human) P07195, (Bovine) Q5E9B1, (Rat) P42123, (Mouse) P16125
Entrez Gene ID: (Dog) 477675, (Human) 3945, (Rabbit) 100009106, (Horse) 100033997, (Bovine) 281275, (Guinea pig) 100714853, (Rat) 24534, (Mouse) 16832
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.