Invitrogen
Manufacturer Code:PA523036
Catalog # PIPA523036
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to an C-terminal portion of the human LDHA protein (within residues 280-332). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell proliferation-inducing gene 19 protein; epididymis secretory sperm binding protein Li 133P; L-lactate dehydrogenase A chain; lactate dehydrogenase 1, A chain; lactate dehydrogenase A4; lactate dehydrogenase M; lactate dehydrogenase-A; LDH muscle subunit; LDH-A; LDH-M; M-LDH; PIG19 GSD11 LDH1 LDHM L-lactate dehydrogenase A chain LDH muscle subunit LDH-A LDH-M cell proliferation-inducing gene 19 protein lactate dehydrogenase M proliferation-inducing gene 19 renal carcinoma antigen NY-REN-59; proliferation-inducing gene 19; Renal carcinoma antigen NY-REN-59
Gene Aliases: GSD11; HEL-S-133P; l7R2; Ldh-1; Ldh1; LDHA; LDHM; PIG19
UniProt ID: (Human) P00338, (Mouse) P06151
Entrez Gene ID: (Human) 3939, (Mouse) 16828
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.