Proteintech
Manufacturer Code:199871AP150UL
Catalog # 50-173-2638
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell proliferation-inducing gene 19 protein; epididymis secretory sperm binding protein Li 133P; L-lactate dehydrogenase A chain; lactate dehydrogenase 1, A chain; lactate dehydrogenase A LDH A LDH M LDH muscle subunit LDH1 LDHA PIG19; lactate dehydrogenase A-like; lactate dehydrogenase A4; lactate dehydrogenase M; lactate dehydrogenase-A; LDH muscle subunit; LDH-A; LDH-M; M-LDH; proliferation-inducing gene 19; Renal carcinoma antigen NY-REN-59
Gene Aliases: GSD11; HEL-S-133P; l7R2; Ldh-1; Ldh1; LDHA; LDHM; PIG19
UniProt ID: (Human) P00338, (Rat) P04642, (Mouse) P06151
Entrez Gene ID: (Human) 3939, (Rat) 24533, (Mouse) 16828
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.