Invitrogen
Manufacturer Code:PA521577
Catalog # PIPA521577
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 86 and 411 of LDB1 (Uniprot IDNo.Q86U70) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: carboxy terminal LIM domain protein 2; Carboxyl-terminal LIM domain-binding protein 2; CLIM-2; CLIM2 NLI CLIM-2 LDB-1 LIM domain-binding factor CLIM2 LIM domain-binding factor-1 LIM domain-binding protein 1 carboxy terminal LIM domain protein 2 carboxyl-terminal LIM domain-binding protein 2 hLdb1 nuclear LIM interactor; hLdb1; LDB-1; LIM domain-binding factor CLIM2; LIM domain-binding factor-1; LIM domain-binding protein 1; mLdb1; Nuclear LIM interactor
Gene Aliases: CLIM-2; CLIM2; LDB-1; LDB1; NLI
UniProt ID: (Human) Q86U70, (Mouse) P70662
Entrez Gene ID: (Human) 8861, (Rat) 309447, (Mouse) 16825
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.