Novus Biologicals
Manufacturer Code:NBP177832
Catalog # NBP177832
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to aa 361-373 of mouse LDB1 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: carboxy terminal LIM domain protein 2; carboxy terminal LIM domain protein 2 Carboxyl-terminal LIM domain-binding protein 2 CLIM-2 CLIM2hLdb1 LDB-1 LIM domain binding 1 LIM domain-binding factor-1 LIM domain-binding protein 1 NLILIM domain-binding factor CLIM2 Nuclear LIM interactor; Carboxyl-terminal LIM domain-binding protein 2; CLIM-2; hLdb1; LDB-1; LIM domain-binding factor CLIM2; LIM domain-binding factor-1; LIM domain-binding protein 1; Nuclear LIM interactor
Gene Aliases: CLIM-2; CLIM2; LDB-1; LDB1; NLI
UniProt ID: (Human) Q86U70
Entrez Gene ID: (Human) 8861
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.