Proteintech
Manufacturer Code:245891AP150UL
Catalog # 50-173-4441
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LCTL Fusion Protein Ag20220 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KL lactase phlorizin hydrolase; KLG KLPH lactase like Lactase like protein LCTL; klotho gamma; Klotho/lactase-phlorizin hydrolase-related protein; lactase-like; Lactase-like protein
Gene Aliases: KLG; KLPH; LCTL; UNQ3022/PRO9820
UniProt ID: (Human) Q6UWM7
Entrez Gene ID: (Human) 197021
Molecular Function: glucosidase glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.