Invitrogen
Manufacturer Code:PA528498
Catalog # PIPA528498
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 193 and 414 of Human LYCAT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-acylglycerol-3-phosphate O-acyltransferase 8; 1-AGP acyltransferase 8; 1-AGPAT 8; 1AGPAT8 AGPAT8 ALCAT1 HSRG1849 LYCAT UNQ1849 1-AGP acyltransferase 8 1-AGPAT 8 1-acylglycerol-3-phosphate O-acyltransferase 8 acyl-CoA:lysocardiolipin acyltransferase 1; Acyl-CoA:lysocardiolipin acyltransferase 1; Lysocardiolipin acyltransferase 1
Gene Aliases: 1AGPAT8; AGPAT8; ALCAT1; HSRG1849; LCLAT1; LYCAT; UNQ1849; UNQ1849/PRO3579
UniProt ID: (Human) Q6UWP7
Entrez Gene ID: (Human) 253558
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.