Invitrogen
Manufacturer Code:PA579587
Catalog # PIPA579587
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human Lck. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Leukocyte C-terminal Src kinase; LSK; Lymphocyte cell-specific protein-tyrosine kinase; lymphocyte protein tyrosine kinase; lymphocyte-specific protein tyrosine kinase; p56(LSTRA) protein-tyrosine kinase; p56-LCK; Protein YT16; Proto-oncogene Lck; Proto-oncogene tyrosine-protein kinase LCK; T cell-specific protein-tyrosine kinase; T-lymphocyte specific protein tyrosine kinase p56lck; Tyrosine-protein kinase Lck; Tyrosine-protein kinase Lck Leukocyte C-terminal Src kinase LSK Lymphocyte cell-specific protein-tyrosine kinase Protein YT16 Proto-oncogene Lck T cell-specific protein-tyrosine kinase p56-LCK LCK
Gene Aliases: Hck-3; IMD22; LCK; Lck1; Lcktkr; LSK; Lsk-t; Lskt; p56<lck>; p56Lck; pp58lck; YT16
UniProt ID: (Human) P06239, (Rat) Q01621, (Mouse) P06240
Entrez Gene ID: (Human) 3932, (Rat) 313050, (Mouse) 16818
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.