R&D Systems
Manufacturer Code:IC9390R100
Catalog # IC9390R100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1251A |
Immunogen | Human LC3B synthetic peptide Accession # Q9GZQ8 |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Apg8b ATG8F Autophagy-related protein LC3 B Autophagy-related ubiquitin-like modifier LC3 B LC3B lc3b autophagy marker lc3-i ii LC3II MAP1 light chain 3-like protein 2 MAP1A/1BLC3 MAP1A/MAP1B LC3 B map1lc3b MAP1LC3B-a microtubule associated protein 1 light chain 3 beta microtubule associated protein 3 b microtubule associated protein 3 beta microtubule-associated proteins 1A/1B light chain 3 microtubule-associated proteins 1A/1B light chain 3B; Autophagy-related protein LC3 B; Autophagy-related ubiquitin-like modifier LC3 B; MAP1 light chain 3-like protein 2; MAP1A/MAP1B LC3 B; MAP1A/MAP1B light chain 3 B; Microtubule-associated protein 1 light chain 3 beta; Microtubule-associated proteins 1A/1B light chain 3B
Gene Aliases: ATG8F; LC3B; MAP1A/1BLC3; MAP1ALC3; MAP1LC3B; MAP1LC3B-a
UniProt ID: (Human) Q9GZQ8
Entrez Gene ID: (Human) 81631
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.