Novus Biologicals
Manufacturer Code:NB1002220SS
Catalog # NB1002220SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunoblot (IB) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Polyclonal LC3B Antibody was made to a synthetic peptide made to an N-terminal portion of the human LC3B protein sequence (between residues 1-100). [UniProt# Q9GZQ8] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: anti-LC3B atg8f Autophagy-related protein LC3 B Autophagy-related ubiquitin-like modifier LC3 B LC3B lc3b autophagy marker LC3B ihc LC3B immunohistochemistry LC3B immunoprecipitation LC3B western blot lc3-i ii MAP1 light chain 3-like protein 2 MAP1A/1BLC3 MAP1A/MAP1B LC3 B map1lc3b MAP1LC3B-a microtubule associated protein 1 light chain 3 beta microtubule associated protein 3 b microtubule associated protein 3 beta microtubule-associated proteins 1A/1B light chain 3 microtubule-associated proteins 1A/1B light chain 3B; Autophagy-related protein LC3 B; Autophagy-related ubiquitin-like modifier LC3 B; MAP1 light chain 3-like protein 2; MAP1A/MAP1B LC3 B; MAP1A/MAP1B light chain 3 B; Microtubule-associated protein 1 light chain 3 beta; Microtubule-associated proteins 1A/1B light chain 3B
Gene Aliases: ATG8F; LC3B; MAP1A/1BLC3; MAP1ALC3; MAP1LC3B; MAP1LC3B-a
UniProt ID: (Human) Q9GZQ8
Entrez Gene ID: (Human) 81631
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.