Proteintech
Manufacturer Code:187251AP150UL
Catalog # 50-173-2211
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Autophagy-related protein LC3 B; Autophagy-related ubiquitin-like modifier LC3 B; LC3 LC3B MAP1A/1BLC3 MAP1A/MAP1B LC3 B MAP1A/MAP1B light chain 3 B MAP1ALC3 MAP1LC3B; MAP1 light chain 3-like protein 2; MAP1A/1B light chain 3 B; MAP1A/MAP1B LC3 B; MAP1A/MAP1B light chain 3 B; Microtubule-associated protein 1 light chain 3 beta; microtubule-associated proteins 1A/1B light chain 3; Microtubule-associated proteins 1A/1B light chain 3B
Gene Aliases: 1010001C15Rik; Atg8; ATG8F; LC3B; MAP1A/1BLC3; MAP1ALC3; Map1lc3; MAP1LC3B; MAP1LC3B-a; Mpl3; zbs559
UniProt ID: (Human) Q9GZQ8, (Rat) Q62625, (Mouse) Q9CQV6
Entrez Gene ID: (Human) 81631, (Rat) 64862, (Mouse) 67443
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.