Invitrogen
Manufacturer Code:MA140030
Catalog # 01-676-488
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RR433-8 |
Immunogen | Murine natural and recombit LBP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: LBP; Lipopolysaccharide binding protein; Lipopolysaccharide-binding protein
Gene Aliases: Bpifd2; Lbp; Ly88
UniProt ID: (Mouse) Q61805
Entrez Gene ID: (Mouse) 16803
Molecular Function:
antibacterial response protein
defense/immunity protein
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.