Invitrogen
Manufacturer Code:MA140129
Catalog # 01-676-520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6G3 |
Immunogen | Human natural and recombit LBP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: acute phase protein; binds bacterial LPS; BPI fold containing family D, member 2; LBP; lipopolysaccharide binding protein; Lipopolysaccharide-binding protein; LPS-binding protein
Gene Aliases: BPIFD2; LBP; LPSBP
UniProt ID: (Bovine) Q2TBI0, (Human) P18428
Entrez Gene ID: (Bovine) 512242, (Pig) 397303, (Rabbit) 100009124, (Human) 3929, (Goat) 102186029, (Dog) 485869, (Sheep) 101104595, (Rat) 29469
Molecular Function:
antibacterial response protein
defense/immunity protein
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.