R&D Systems
Manufacturer Code:MAB8701SP
Catalog # MAB8701SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 127420 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human LBP Ala26-Val481 Accession # AAD21962 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
Protein Aliases: BPI fold containing family D, member 2; LBP; lipopolysaccharide binding protein lipopolysaccharide-binding protein LPS-binding protein MGC22233; Lipopolysaccharide-binding protein; LPS-binding protein
Gene Aliases: BPIFD2; LBP
UniProt ID: (Human) P18428
Entrez Gene ID: (Human) 3929
Molecular Function:
antibacterial response protein
defense/immunity protein
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.