Invitrogen
Manufacturer Code:PA579586
Catalog # PIPA579586
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived mouse LBP recombit protein (Position: V26-R257). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BPI fold containing family D, member 2; LBP; Lipopolysaccharide-binding protein; Lipopolysaccharide-binding protein LBP Lbp
Gene Aliases: Bpifd2; Lbp; Ly88
UniProt ID: (Mouse) Q61805
Entrez Gene ID: (Mouse) 16803, (Rat) 29469
Molecular Function:
antibacterial response protein
defense/immunity protein
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.