Novus Biologicals
Manufacturer Code:NBP159431
Catalog # NBP159431
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLC43A1(solute carrier family 43 member 1) The peptide sequence was selected from the middle region of SLC43A1. Peptide sequence AVNKMLEYLVTGGQEHETNEQQQKVAETVGFYSSVFGAMQLLCLLTCPLI. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: L-type amino acid transporter 3; Large neutral amino acids transporter small subunit 3; LAT3prostate cancer overexpressed gene 1 PB39L-type amino acid transporter 3 POV1large neutral amino acids transporter small subunit 3 Prostate cancer overexpressed gene 1 protein R00504 Solute carrier family 43 member 1 solute carrier family 43 member 1; Prostate cancer overexpressed gene 1 protein; SLC43A1; solute carrier family 43 (amino acid system L transporter), member 1; Solute carrier family 43 member 1; solute carrier family 43, member 1
Gene Aliases: LAT3; PB39; POV1; R00504; SLC43A1
UniProt ID: (Human) O75387
Entrez Gene ID: (Human) 8501
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.