Invitrogen
Manufacturer Code:PA520648
Catalog # PIPA520648
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 13 amino acid synthetic peptide near the carboxy terminus of the human LASS6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ceramide synthase 6; CerS5 LASS6 LAG1 homolog ceramide synthase 6 LAG1 longevity assurance homolog 6 longevity assurance homolog 6; CerS6; LAG1 homolog, ceramide synthase 6; LAG1 longevity assurance homolog 6; longevity assurance homolog 6
Gene Aliases: 4732462C07Rik; AW544719; CERS5; CERS6; LASS6; RGD1561599; T1L
UniProt ID: (Human) Q6ZMG9, (Mouse) Q8C172
Entrez Gene ID: (Human) 253782, (Mouse) 241447, (Rat) 366065
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.