Novus Biologicals
Manufacturer Code:NBP256406
Catalog # NBP256406
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:WLPHNVTWADLKNTEEATFPQAEDL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ceramide synthase 6; CerS6; CerS6 LAG1 homolog ceramide synthase 6 LAG1 longevity assurance homolog 6 LAG1 longevity assurance homolog 6 (S. cerevisiae) longevity assurance homolog 6 MGC129949 MGC129950; LAG1 homolog, ceramide synthase 6; LAG1 longevity assurance homolog 6; longevity assurance homolog 6
Gene Aliases: CERS5; CERS6; LASS6
UniProt ID: (Human) Q6ZMG9
Entrez Gene ID: (Human) 253782
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.