Invitrogen
Manufacturer Code:PA520647
Catalog # PIPA520647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 14 amino acid peptide near the amino terminus of the human LASS5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ceramide synthase 5; CerS5; LAG1 homolog, ceramide synthase 5; LAG1 longevity assurance homolog 5; LASS5 Trh4 LAG1 homolog ceramide synthase 5 LAG1 longevity assurance homolog 5; longevity assurance homolog 5; Sphingosine N-acyltransferase CERS5; TRAM homolog 4; Translocating chain-associating membrane protein homolog 4
Gene Aliases: 2310081H14Rik; AU045339; AW544927; CERS5; LASS5; Trh4
UniProt ID: (Human) Q8N5B7, (Mouse) Q9D6K9
Entrez Gene ID: (Human) 91012, (Mouse) 71949, (Rat) 366984
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.