Abnova
Manufacturer Code:PAB13439
Catalog # 89-116-681
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus 17 amino acids of human LASS5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of LASS5.
For Research Use Only
Protein Aliases: Ceramide synthase 5; CerS5; FLJ25304; LAG1 homolog, ceramide synthase 5; LAG1 longevity assurance homolog 5; MGC45411; Sphingosine N-acyltransferase CERS5; Trh4
Gene Aliases: CERS5; LASS5; Trh4
UniProt ID: (Human) Q8N5B7
Entrez Gene ID: (Human) 91012
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.