Abnova
Manufacturer Code:H00029956M11A
Catalog # 89-106-803
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3G15 |
Immunogen | LASS2 (AAH013571 a.a. &bsim 380 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant LASS2.
For Research Use Only
Protein Aliases: Ceramide synthase 2; CerS2; FLJ10243; L3; LAG1 homolog, ceramide synthase 2; LAG1 longevity assurance 2; LAG1 longevity assurance homolog 2; longevity assurance (LAG1, S. cerevisiae) homolog 2; MGC987; SP260; Sphingosine N-acyltransferase CERS2; TMSG1; Tumor metastasis-suppressor gene 1 protein
Gene Aliases: CERS2; L3; LASS2; SP260; TMSG1
UniProt ID: (Human) Q96G23
Entrez Gene ID: (Human) 29956
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.