Proteintech
Manufacturer Code:160101AP150UL
Catalog # 50-173-0639
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LAS1L Fusion Protein Ag8732 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LAS1 like (S. cerevisiae) LAS1L Protein LAS1 homolog; LAS1-like, ribosome biogenesis factor; Protein LAS1 homolog; Ribosomal biogenesis protein LAS1L
Gene Aliases: 1810030A06Rik; 5830482G23Rik; dJ475B7.2; Las1-like; LAS1L; MSTP060; RGD1565755
UniProt ID: (Human) Q9Y4W2, (Mouse) A2BE28
Entrez Gene ID: (Human) 81887, (Mouse) 76130, (Rat) 296865
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.