Invitrogen
Manufacturer Code:PA595627
Catalog # PIPA595627
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human Leucyl tRNA synthetase/LARS recombit protein (Q628-Q855). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cytoplasmic leucyl-tRNA synthetase; cytosolic leucyl-tRNA synthetase; leucine translase; leucine tRNA ligase 1, cytoplasmic; Leucine--tRNA ligase cytoplasmic Leucyl-tRNA synthetase LeuRS LARS KIAA1352; Leucine--tRNA ligase, cytoplasmic; Leucyl-tRNA synthetase; leucyl-tRNA synthetase, cytoplasmic; LeuRS; proliferation-inducing gene 44
Gene Aliases: 2310045K21Rik; 3110009L02Rik; AW536573; hr025Cl; HSPC192; ILFS1; KIAA1352; LARS; LARS1; LEURS; LEUS; LFIS; LRS; mKIAA1352; PIG44; RNTLS
UniProt ID: (Human) Q9P2J5, (Mouse) Q8BMJ2
Entrez Gene ID: (Human) 51520, (Mouse) 107045, (Rat) 291624
Molecular Function: aminoacyl-tRNA synthetase ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.