Invitrogen
Manufacturer Code:PA571490
Catalog # PIPA571490
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 176-205 amino acids from the N-terminal region of human LARS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSPC192 LARS1 LEURS LEUS LRS PIG44 RNTLS hr025Cl cytoplasmic leucyl-tRNA synthetase cytosolic leucyl-tRNA synthetase leucine tRNA ligase 1 cytoplasmic leucine translase leucine--tRNA ligase cytoplasmic leucine-tRNA ligase leucyl-tRNA synthetase cytoplasmic proliferation-inducing gene 44; Leucine--tRNA ligase, cytoplasmic; Leucyl-tRNA synthetase; leucyl-tRNA synthetase, cytoplasmic; LeuRS
Gene Aliases: 2310045K21Rik; 3110009L02Rik; AW536573; Lars; Lars1; mKIAA1352
UniProt ID: (Mouse) Q8BMJ2
Entrez Gene ID: (Mouse) 107045
Molecular Function:
aminoacyl-tRNA synthetase
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.