Invitrogen
Manufacturer Code:PA559338
Catalog # PIPA559338
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human LARS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cytoplasmic leucyl-tRNA synthetase; cytosolic leucyl-tRNA synthetase; HSPC192 LARS1 LEURS LEUS LRS PIG44 RNTLS hr025Cl cytoplasmic leucyl-tRNA synthetase cytosolic leucyl-tRNA synthetase leucine tRNA ligase 1 cytoplasmic leucine translase leucine--tRNA ligase cytoplasmic leucine-tRNA ligase leucyl-tRNA synthetase cytoplasmic proliferation-inducing gene 44; leucine translase; leucine tRNA ligase 1, cytoplasmic; Leucine--tRNA ligase, cytoplasmic; Leucyl-tRNA synthetase; LeuRS; proliferation-inducing gene 44
Gene Aliases: hr025Cl; HSPC192; ILFS1; KIAA1352; LARS; LARS1; LEURS; LEUS; LFIS; LRS; PIG44; RNTLS
UniProt ID: (Human) Q9P2J5
Entrez Gene ID: (Human) 51520
Molecular Function: aminoacyl-tRNA synthetase ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.