Invitrogen
Manufacturer Code:PA514309
Catalog # PIPA514309
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1118-1145 amino acids from the C-terminal region of human LARS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cytoplasmic leucyl-tRNA synthetase; cytosolic leucyl-tRNA synthetase; leucine translase; leucine tRNA ligase 1, cytoplasmic; Leucine--tRNA ligase, cytoplasmic; Leucyl-tRNA synthetase; Leucyl-tRNA synthetase cytoplasmic Leucine--tRNA ligase LeuRS LARS; LeuRS; proliferation-inducing gene 44
Gene Aliases: hr025Cl; HSPC192; ILFS1; KIAA1352; LARS; LARS1; LEURS; LEUS; LFIS; LRS; PIG44; RNTLS
UniProt ID: (Human) Q9P2J5
Entrez Gene ID: (Human) 51520
Molecular Function:
aminoacyl-tRNA synthetase
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.