Invitrogen
Manufacturer Code:PA578393
Catalog # PIPA578393
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human LARGE. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylglucosaminyltransferase-like 1A; acetylglucosaminyltransferase-like protein; Beta-1,3-glucuronyltransferase LARGE; Glycosyltransferase-like protein; glycosyltransferase-like protein LARGE1; LARGE xylosyl- and glucuronyltransferase 1; like-glycosyltransferase; MDDGB6 MDC1D MDDGA6 like-glycosyltransferase; myodystrophy; Xylosyltransferase LARGE
Gene Aliases: BPFD#36; enr; fg; froggy; Gyltl1a; KIAA0609; LARGE; LARGE1; Mbp-1; Mbp1; MDC1D; MDDGA6; MDDGB6; mKIAA0609; myd
UniProt ID: (Human) O95461, (Mouse) Q9Z1M7
Entrez Gene ID: (Human) 9215, (Mouse) 16795, (Rat) 361368
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.