Invitrogen
Manufacturer Code:PA542891
Catalog # PIPA542891
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human LARGE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylglucosaminyltransferase-like 1A; acetylglucosaminyltransferase-like protein; Beta-1,3-glucuronyltransferase LARGE; Eosinophil granule major basic protein 1; glycosyltransferase-like 1A; Glycosyltransferase-like protein; glycosyltransferase-like protein LARGE1; large; LARGE xylosyl- and glucuronyltransferase 1; like-glycosyltransferase; Like-glycosyltransferase KIAA0609 MDC1D MDDGA6 MDDGB6; MBP-1; myodystrophy; prepro major basic protein 1; Xylosyltransferase LARGE
Gene Aliases: BPFD#36; enr; fg; froggy; Gyltl1a; KIAA0609; LARGE; LARGE1; Mbp-1; MBP1; MDC1D; MDDGA6; MDDGB6; mKIAA0609; myd
UniProt ID: (Human) O95461, (Mouse) Q9Z1M7, (Guinea pig) P22032, (Zebrafish) Q66PG2
Entrez Gene ID: (Horse) 100068935, (Dog) 474521, (Human) 9215, (Rabbit) 100346008, (Bovine) 506466, (Mouse) 16795, (Rat) 361368, (Guinea pig) 100286788, (Guinea pig) 100736069, (Zebrafish) 446213
Molecular Function:
glycosyltransferase
transferase
antibacterial response protein
defense/immunity protein
extracellular matrix protein
extracellular matrix structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.