Invitrogen
Manufacturer Code:PA519292
Catalog # PIPA519292
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (SEADVNSENLQKQ) corresponding to the internal amino acids of LARGE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylglucosaminyltransferase-like 1A; acetylglucosaminyltransferase-like protein; Beta-1,3-glucuronyltransferase LARGE; Glycosyltransferase-like protein; LARGE like-glycosyltransferase KIAA0609 MDC1D acetylglucosaminyltransferase-like 1A acetylglucosaminyltransferase-like protein like-acetylglucosaminyltransferase; LARGE xylosyl- and glucuronyltransferase 1; like-glycosyltransferase; Xylosyltransferase LARGE
Gene Aliases: KIAA0609; LARGE; LARGE1; MDC1D; MDDGA6; MDDGB6
UniProt ID: (Human) O95461
Entrez Gene ID: (Human) 9215
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.