Invitrogen
Manufacturer Code:PA567016
Catalog # PIPA567016
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human LAPTM4B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: lysosomal associated protein transmembrane 4 beta; Lysosomal protein Transmembrane 4 beta LAPTM4beta LC27; Lysosomal-associated transmembrane protein 4B; Lysosome-associated transmembrane protein 4-beta
Gene Aliases: LAPTM4B; LAPTM4beta; LC27; PSEC0001
UniProt ID: (Human) Q86VI4
Entrez Gene ID: (Human) 55353
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.