Invitrogen
Manufacturer Code:PA543047
Catalog # PIPA543047
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human LAPTM4B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: lysosomal associated protein transmembrane 4 beta; Lysosomal protein Transmembrane 4 beta LAPTM4beta LC27; lysosomal-associated protein transmembrane 4B; lysosomal-associated transmembrane protein 4; Lysosomal-associated transmembrane protein 4B; Lysosomal-associated transmembrane protein 4B-like protein; Lysosome-associated transmembrane protein 4-beta
Gene Aliases: C330023P13Rik; LAPTM4B; LAPTM4beta; LC27; PSEC0001
UniProt ID: (Human) Q86VI4, (Bovine) Q71SV0, (Rat) Q5U1W4, (Mouse) Q91XQ6
Entrez Gene ID: (Human) 55353, (Bovine) 404155, (Dog) 475042, (Horse) 100059945, (Rabbit) 100356277, (Rat) 315047, (Mouse) 114128, (Guinea pig) 100730523
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.